Learn More
Enzo Life Sciences Green fluorescent protein polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ALX210199R100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 26-39 (H26KFSVSGEGEGDAT39) of the jellyfish Aequorea victoria GFP (green fluorescent protein). This sequence is completely conserved between GFP and the modified and enhanced Blue, Green and Yellow FP variants. Species reactivity: Species independent. Specificity: Recognizes GFP and GFP fusion proteins. Applications: ICC, WB. Purity: Epitope-affinity purified. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.05% sodium azide.
Catalog No. 50-200-8089
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.