Learn More
IBA LifeSciences GFP-Strep-tag control protein Strep-tagII; 500 µg
Supplier:  IBA LifeSciences 21006005
Encompass_Preferred
GFP-Strep-tagII is a control protein consisting of the green fluorescent protein (GFP) C-terminally tagged with the Strep-tagII. It is suitable for test purifications, detections, and immobilizations with Strep-Tactin or Strep-TactinXT.
Catalog No. 50-210-0129
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.