Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GFR alpha-2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01670650
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-650 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-650 Supplier Invitrogen™ Supplier No. PA118399
  • $617.00
    $497.30 / Each of 1

    Save $119.70

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Glycerol (1:1) may be added for an additional stability. Avoid repetitive freeze/thaw cycles.

GFRa-2 binds NTN and mediates activation of RET receptor tyrosine kinase by both NTN and GDNF. Thus, NTN, GFRa-2, and the Ret PTK form a complex to transduce NTN signal and to mediate NTN function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFR alpha-2
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with no preservative
Gene GFRA2
Gene Accession No. O00451
Gene Alias GDNF family receptor alpha 2; GDNF family receptor alpha-2; GDNF re; GDNF receptor alpha-2; GDNF receptor beta; GDNFR-alpha-2; Gdnfrb; GDNFR-beta; Gfra2; GFR-alpha 2; GFR-alpha-2; glial cell line derived neurotrophic factor family receptor alpha 2; glial cell line derived neurotrophic factor receptor, beta; HpV18gp1; neurturin receptor alpha; NRTNR-alpha; NTNRA; NTNR-alpha; PI-linked cell-surface accessory protein; RET ligand 2; RETL2; TGF-beta-related neurotrophic factor receptor 2; TRN receptor, GPI-anchored; TRNR2; tyrosine kinase receptor ligand 2
Gene Symbols GFRA2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues P(376) R V E K T P S L P D D L S D(390) of human GFR alpha-2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25136, 2675
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.