Learn More
Invitrogen™ GFR alpha-3 Polyclonal Antibody

Description
Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Glycerol (1:1) may be added for an additional stability. Avoid repetitive freeze/thaw cycles.
Specifications
Specifications
| Antigen | GFR alpha-3 |
| Applications | Immunohistochemistry (Frozen), Western Blot |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | whole serum with no preservative |
| Gene | Gfra3 |
| Gene Accession No. | O35118 |
| Gene Alias | GDNF family receptor alpha 3; GDNF family receptor alpha-3; GDNF receptor alpha-3; GDNF-family receptor alpha 3; GDNFR3; GDNFR-alpha-3; GFR alpha-3; GFRA3; GFRalpha3; GFR-alpha-3; glial cell line derived neurotrophic factor family receptor alpha 3; glial cell line-derived neurotrophic factor receptor alpha-3; GPI-linked receptor; UNQ339/PRO538/PRO3664; Y15110 |
| Gene Symbols | Gfra3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.