Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GFR alpha-4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104239
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104239 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104239 Supplier Invitrogen™ Supplier No. PA5104239
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GFRA4.

The protein encoded by this gene is a member of the GDNF receptor family. It is a glycosylphosphatidylinositol(GPI)-linked cell surface receptor for persephin, and mediates activation of the RET tyrosine kinase receptor. This gene is a candidate gene for RET-associated diseases. Alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GFR alpha-4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Gfra4
Gene Accession No. Q9GZZ7
Gene Alias AW060733; G630015H18Rik; GDNF family receptor alpha 4; GDNF family receptor alpha-4; GDNF R alpha-4; GDNF receptor alpha-4; GDNFR-alpha-4; GFR alpha-4; GFR receptor alpha 4; GFRA4; GFR-alpha-4; glial cell line derived neurotrophic factor family receptor alpha 4; neurotrophic factor receptor splice variant A (Gfra4 gene); persephin receptor
Gene Symbols Gfra4
Host Species Rabbit
Immunogen A synthesized peptide derived from human GFRA4(Accession Q9GZZ7), corresponding to amino acid residues L147-A197.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64096
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.