Learn More
Hardy Diagnostics™ QuickSlide™ GramPRO 1 Automated Gram Stain Instrument Control Slides
Catalog No. 23001677 Shop All Fisherbrand Products
Change view
Click to view available options
50 Slides
Catalog No.
23-001-677 50 Slides
1 options
This item is not returnable. View return policy
Catalog No. 23-001-677
This product is either new or inactive, and cannot be ordered online.
This item is not returnable. View return policy
Prepared from standard stock ATCC quality control organisms. Hardy Diagnostics™ QuickSlide™ GramPRO 1 Automated Gram Stain Instrument Control Slides are intended to be used in conjunction with the GramPRO 1 Automatic Gram Stainer (AGS-1000),manual staining.
  • Prepared from standard stock ATCC quality control organisms
  • Gram positive organism: Staphylococcus aureus
  • Gram negative organism: E.coli
  • Used in conjunction with the GramPRO 1 Automatic Gram Stainer (AGS-1000), also for manual staining
  • 50 slides/ Pk.