Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GGCX Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7207062 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-0792 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-0792 Supplier Proteintech Group Inc Supplier No. 162091AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GGCX (gamma-glutamyl carboxylase), also known as GC or VKCFD1 (Vitamin K-dependent gamma-carboxylase), is a 758 amino acid multi-pass membrane protein. Localized to the membrane of the endoplasmic reticulum, GGCX functions to mediate the vitamin K-dependent carboxylation of glutamate residues on target proteins, thereby producing calcium binding gamma-carboxyglutamate (Gla) residues on these proteins and simultaneously converting vitamin K to vitamin K epoxide. GGCX exists as a monomer and, via its ability to modify glutamate residues, it accomplishes the post-translational changes that are necessary for the activity of all vitamin K-dependent proteins (such as blood coagulation and bone matrix proteins). Defects in the gene encoding GGCX are the cause of combined deficiency of vitamin K-dependent clotting factors 1 (VKCFD1) and PXE-like disorder with multiple coagulation factor deficiency, both of which are characterized by abnormal skin, blood or bone function.

Specifications

Antigen GGCX
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GGCX
Gene Accession No. O88496, P38435, Q9QYC7
Gene Alias gamma glutamyl carboxylase, GC, GGCX, VKCFD1
Gene Symbols Ggcx
Host Species Rabbit
Immunogen GGCX Fusion Protein Ag9185
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2677, 56316, 81716
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.