Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GGH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101599
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5101599 100 μL
1 options

Catalog No. PIPA5101599

Supplier: Invitrogen™ PA5101599

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GGH.

Gamma-Glutamyl hydrolase (GGH) is a homodimeric protein that catalyzes the cleavage of the gamma-glutamyl chain of folylpoly-gamma-glutamyl substrates and is a central enzyme in folyl and antifolyl poly-gamma-glutamate metabolism. The activities of the baculovirus-expressed enzyme and the enzyme from HL-60 cells were similar on methotrexate polyglutamates. GGH is expressed in a variety of human tumors and correlates with poor prognosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GGH
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Ggh
Gene Accession No. Q92820, Q9Z0L8
Gene Alias conjugase; FGPH; folylpolyglutamate hydrolase; gamma-GH; gamma-glutamyl hydrolase; gamma-glutamyl hydrolase (conjugase, folylpolygammaglutamyl hydrolase); Gamma-Glu-X carboxypeptidase; GGH; GH
Gene Symbols Ggh
Host Species Rabbit
Immunogen A synthesized peptide derived from human GGH(Accession Q92820), corresponding to amino acid residues L255-T305.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14590, 8836
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.