Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GGT1 Monoclonal Antibody (C1)
GREENER_CHOICE

Catalog No. MA566058
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA566058 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA566058 Supplier Invitrogen™ Supplier No. MA566058
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

GGT1 Monoclonal Antibody for Western Blot, IHC (P)

Glutamyltransferase (gGT) from human kidney is a membrane-bound enzyme that transfers the gamma-glutamly moiety of gamma-glutamyl compounds such as glutathione to an acceptor, which may be an amino acid or a di- or tripeptide or water. The principal functions of gGT may be hydrolysis and metabolism of glutathione. Various human gGT isoforms have been found. The molecular mass of gGT for normal kidney is 90kDa and gGT can be used as a renal cell marker.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GGT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone C1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene GGT1
Gene Accession No. P19440, P20735
Gene Alias CD224; D22S672; D22S732; dwg; gamma glutamyl-transferase 1; gamma-glutamyl transpeptidase; gamma-glutamyltransferase 1; Gamma-glutamyltranspeptidase 1; GGLUT; Ggt; GGT 1; Ggt1; GGT-1; Ggtp; glutathione hydrolase 1; Glutathione hydrolase 1 heavy chain; Glutathione hydrolase 1 light chain; Glutathione hydrolase 1 proenzyme; GTG; Leukotriene-C4 hydrolase; testicular tissue protein Li 73
Gene Symbols GGT1
Host Species Mouse
Immunogen Recombinant Gamma-Glutamyltransferase 1 (gGT1) (Ser122-Ser299).
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2678, 397095
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.