Learn More
Beckman Coulter GH 3.8 HORIZONTAL ROTOR ASSEMB
Supplier:  Beckman Coulter 360581
Encompass_Preferred
GH 3.8 Horizontal Rotor Assembly~UIncludes~U: ~B Rotor ~B Yoke ~B Fourmachined aluminum buckets ~B 3,750 rpmmaximum speed ~B 3,200g maximum force~B 3 liter capacity ~B Aerosol containmentcapabilities ~B Accepts all modular diskadapters ~B 7 year rotor warranty ~B Accepts microplate carrier accessory
Catalog No. NC2110084
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.