Learn More
Enzo Life Sciences Ghrelin (human) (0.5mg). CAS: 258279-04-8
SDP
Supplier:  Enzo Life Sciences ALX157021MC05
Encompass_Preferred
Ghrelin, a 28 aa peptide predominantly produced by the stomach, is an endogenous ligand for the growth hormone secretagogue receptor (GHS-R) that potently stimulates growth hormone release. Studies suggest that peripherally or centrally administrated ghrelin, independent of growth hormone, decreases fat oxidation and increases food intake and adiposity. In addition, plasma ghrelin levels are lower in obese human subjects. Ghrelin promotes the production of orexigenic neuropeptides (neuropeptide Y and agouti-related protein) in the hypothalamic arcuate nuclei and activates the neurons which produce these orexigenic neuropeptides, resulting in an increase in feeding and body weight and thereby opposing the effects of leptin. Moreover, ghrelin shows regulatory effects on the cardiovascular and immune system. Formula: C149H249N47O42. MW: 3370.9. Purity: ≥98%. Formulation: Lyophilized. Solubility: Soluble in distilled water or 5% acetic acid. Long Term Storage: -20°C.
Catalog No. 50-200-7823
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.