Learn More
Abcam Ghrelin (human), Endogenous GHS Agonist, Solid, 1 mg
Description
- Molecular weight: 3,370.91 g/mol
- Purity: >95%
- Form: Solid
- Activity: Endogenous agonist peptide for the ghrelin (GHS) receptor
- Biological effect: Stimulates release of growth hormone from the pituitary gland and regulates feeding, appetite, growth and energy homeostasis
- Storage: Store at -20°C
- Shipping: Ambient - Can Ship with Ice
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Reactivity | Human |
| Shipping Condition | Ambient |
| Target Molecule | Ghrelin Receptor |
| Formulation | 100% Ghrelin (human) |
| Quantity | 1 mg |
| For Use With (Application) | Ghrelin (GHS) receptor stimulation, Growth hormone release, Feeding regulation, Appetite regulation, Growth regulation, Energy homeostasis regulation |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.