Learn More
Bachem Ghrelin (mouse, rat) Trifluoroacetate
SDP
Supplier:  Bachem 4033076.0500
Encompass_Preferred
Ghrelin (mouse, rat) Trifluoroacetate, H-Gly-Ser-Ser(octanoyl)-Phe-Leu-Ser-Pro-Glu-His-Gln-Lys-Ala-Gln-Gln-Arg-Lys-Glu-Ser-Lys-Lys-Pro-Pro-Ala-Lys-Leu-Gln-Pro-Arg-OH, 258338-12-4, C147H245N45O42, Mr 3314.84, 0.5mg. Ghrelin, a peptide hormone produced by the stomach oxyntic cells plays a crucial role in appetite regulation. It binds to the growth hormone secretagogue receptor (GHS-R), which stimulates the release of GH. Ghrelin, which promotes food uptake and body weight increase, acts as an antagonist of leptin. Thus, it has become an important tool in obesity research. Additionally, ghrelin is involved in the bone metabolism, in reproduction, and in the immune system.
Catalog No. 50-259-884
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.