Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Ghrelin Polyclonal Antibody
Chicken Polyclonal Antibody
Supplier: Invitrogen™ PA19595
Description
PA1-9595 detects Ghrelin in human and mouse samples. PA1-9595 has been successfully used in ELISA procedures. PA1-9595 immunogen is a synthetic active human ghrelin peptide.
GHRL encodes ghrelin-obestatin preproprotein, which generates ghrelin and obestatin. Ghrelin is an the ligand for the growth hormone secretagogue receptor type 1 and is involved in regulating growth hormone release. Ghrelin has an appeptite-stimulating effect and induces adiposity and stimulates gastric acid secretion. Obestatin may be the ligand for the orphan G protein-coupled receptor GPR39 and is involved in satiety and can have an appetite-reducing effect resulting in decreased food intake.
Specifications
Ghrelin | |
Polyclonal | |
Unconjugated | |
GHRL | |
2210006E23Rik; AMCBX1; Appetite-regulating hormone; appetite-regulating hormone; uncharacterized protein LOC58991; exon 2- and 3-deleted preproghrelin; exon 2-deleted preproghrelin; FIP3; FIP-3; Fip3p; Ghr; Ghrelin; ghrelin 27; ghrelin 28; ghrelin and obestatin prepropeptide; ghrelin precursor; ghrelin, growth hormone secretagogue receptor ligand; ghrelin/obestatin; ghrelin/obestatin preprohormone; ghrelin/obestatin prepropeptide; Ghrelin-27; Ghrelin-28; ghrelin-like protein; GHRL; Growth hormone secretagogue; growth hormone-releasing peptide; IKK-gamma; In2c-preproghrelin; IP; IP1; IP2; IPD2; m46; Motilin-related peptide; MTLRP; MTLRPAP; NEMO; Obestatin; Obestatin-13; Obestatin-23; OTTHUMP00000207794; OTTHUMP00000207795; OTTHUMP00000207796; OTTHUMP00000207798; prepro-appetite regulatory hormone; preproghrelin; proghrelin; Protein M46; UNQ524/PRO1066 | |
Chicken | |
Affinity chromatography | |
RUO | |
51738, 58991 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9EQX0, Q9UBU3 | |
GHRL | |
Synthetic active human ghrelin peptide. | |
200 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgY |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction