Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Ghrelin (Rat), Endogenous GHS Agonist, Solid, >95% Purity, 1 mg

Catalog No. 502707774
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7774 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7774 Supplier Abcam Supplier No. AB1202311MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Highly pure endogenous growth hormone secretagogue agonist for physiological studies.

  • Molecular Weight: 3314.8 Da
  • Purity: >95%
  • Form: Solid
  • Storage Temperature: Store at -20°C
  • Shipping Temperature: Ambient - Can Ship with Ice
  • Composition: 100% Ghrelin (rat)
  • CAS Number: 258338-12-4

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Ghrelin (Rat)
Shipping Condition Ambient
Substrate Properties Peptide-based Substrate
Form Solid
Content And Storage Store at -20°C.
Quantity 1 mg
For Use With (Application) Functional Studies
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.