Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GHRHR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA3117 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA3117 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA3117 Supplier Invitrogen™ Supplier No. PA3117
  • $567.00
    $428.65 / Each of 1

    Save $138.35

    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA3-117 detects GHRH-R in human samples. PA3-117 has been successfully used in Western blot, immunocytochemistry and immunohistochemistry procedures. By Western blot, this antibody detects an ~40 kDa protein representing GHRH-R in human samples. In immunohistochemistry, GHRH-R immunoreactivity is predominantly localized at the plasma membrane. The PA3-117 immunizing peptide corresponds to amino acid residues 403-422 from human GHRH-R.

Growth Hormone Releasing Hormone (GHRH) is a member of a family of peptide hormones that also includes VIP (vasoactive intestinal polypeptide) and PACAP (pituitary adenylate cyclase-activating polypeptide). It is a hypothalamic peptide that binds to G-protein coupled receptors to stimulate secretion and synthesis of growth hormone (GH). GHRH receptor proteins have been identified on the plasma membrane of primary human tumor cells, and are thought to be involved in the growth of a variety of cancer cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GHRHR
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.05% sodium azide
Gene Ghrhr
Gene Accession No. Q02643
Gene Alias GHRFR; GHRH receptor; GHRHR; Ghrh-r; GHRHREC; GHRHRpsv; GRF receptor; GRFR; Growth hormone - releasing receptor; growth hormone releasing hormone receptor; growth hormone-releasing factor receptor; Growth hormone-releasing hormone receptor; IGHD1B; lit; little
Gene Symbols Ghrhr
Host Species Rabbit
Immunogen Synthetic peptide, KLH conjugated, corresponding to residues R(403) T R A K W T T P S R S A A K V L T S M(422) of human GHRH-R.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2692
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.