Learn More
HCL Labels Pre-Labeled GHS Spray Bottles - Custom (Send in your SDS) - 16oz
SDP
Supplier:  HCL Labels GHSBOTXX16
Encompass_Preferred
This item has a minimum qty of 5 packs per supplier requirements.
GHS Compliant - Pre-labeled Spray Bottles - 16 oz - These bottles include a pre-affixed GHS hazard communication label, and are ready to be filled (The bottles are empty)Made from chemical resistant HDPE material including a reinforced sprayer - they will not fail, or leak - even after heavy use.Send in your SDS for your custom chemical.Bottles include caps for when a sprayer isn't needed.Bottles have a fully adjustable spray nozzle - 1.4ml per strokeLabels are made of a durable Adhesive Vinyl that is laminated for chemical and solvent resistance.
Catalog No. NC1974415
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.