Learn More
Apexbio Technology LLC GI 254023X 260264-93-5 5mg
SDP
Supplier:  Apexbio Technology LLC A44365
Encompass_Preferred
GI 254023X (CAS 260264-93-5) is a selective small-molecule inhibitor targeting ADAM10 (a disintegrin and metalloprotease 10) a membrane-associated protease involved in constitutive ectodomain shedding of diverse cell-surface proteins This compound exhibits potent inhibition against ADAM10 with an IC50 of approximately 5 3 nM and displays a selectivity profile over ADAM17 (TACE) by more than 100-fold Research studies have demonstrated that GI 254023X effectively reduces constitutive cleavage of fractalkine (CX3CL1) in ECV-304 transfectants in vitro Moreover in a mouse model of vascular injury induced by Staphylococcus aureus -hemolysin GI 254023X treatment was associated with delayed progression of endothelial damage Currently GI 254023X remains at the preclinical research stage
Catalog No. 50-000-61632
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.