Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ CD Hybridoma AGT™ Medium

Catalog No. 12372001
Encompass
Change view
Click to view available options
Quantity:
1 L
10 L
100 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-372-001 100 L
12-372-025 1 L
12-372-017 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-372-001 Supplier Gibco™ Supplier No. 12372001
Add to Cart
Edge
Add to Cart
Request a Quote

Animal origin-free, chemically defined, protein-free medium

Gibco CD Hybridoma Medium is specifically developed for the ability to support the growth of hybridomas for antibody production.

  • Ability to support suspension and stationary hybridomas and myelomas
  • Formulated without L-glutamine for stability
  • Animal origin-free, protein-free and chemically defined formulation
  • Convenient, easy-to-use Advanced Granulation Technology (AGT™) format

Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Wet ice

Specifications

Cell Type Myeloma Cells, Hybridomas
Classification Animal Origin-free, Chemically-defined, Protein-free
Concentration 1 X
Form Powder (AGT)
Product Type Hybridoma Medium
Serum Level Serum-free
Sterility Non-sterile
With Additives Sodium Pyruvate, Sodium Bicarbonate
Without Additives No Glutamine, No HEPES, No Phenol Red
Glucose Concentration 3.66 g/L
Product Line AGT, Gibco
Quantity 100 L
Shipping Condition Wet Ice
Endotoxin Level Low
Inorganic Salts Calcium, Magnesium, Sodium Phosphate
Purity or Quality Grade Certified
Content And Storage Storage conditions: 2-8°C, dark and dry
Shipping conditions: Gel packs
Shelf life: 24 months from date of manufacture
Culture Environment CO2
Show More Show Less
What are the main benefits of using AGT?

Complete formulations
- Homogeneous distribution of minute components
- Complete complex formulations in a dry format, including serum-free, protein-free, and chemically defined nutrient media
- Pre-adjusted to the appropriate pH and osmolality
- Require only standard supplementation with L-glutamine for L-glutamine dependent systems
- Can be customized for modified catalog and customers' proprietary formulations

Granular format
- Dissolves instantly for faster media preparation times
- Dust-free granules reduce mess and environmental contamination
- Performance equivalent to liquid media
- Workflow improvement
- Helps reduce total cycle costs due to lower number of components, decreasing time involved in raw material planning, procurement, and testing
- Faster and easier media preparation, with less potential for error
- Easy to scale up from research through production, in batch sizes from 2 kg to 6,000 kg
- Available in various packaging options

What is AGT?

Granulated cell culture media is an easy-to-use, granular form of dry media that is produced by applying fluid bed granulation technology to cell culture nutrient media manufacturing. The patented process allows production of complete formulations of a variety of serum-free, protein-free, and chemically defined media in a dry format.

AGT offers the benefits of liquid media without the cost, storage, and transportation issues. It is a complete medium that is pH and osmolality pre-adjusted. The granules dissolve instantly for faster media preparation time than conventional dry powder media. AGT media can therefore help reduce total cycle costs, decreasing time involved in raw material planning, procurement, and testing as well as media preparation. These benefits are all realized while maintaining comparable cell growth to liquid media.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.