Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Soybean Trypsin Inhibitor, powder

Catalog No. 17075029
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-075-029 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-075-029 Supplier Gibco™ Supplier No. 17075029
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trypsin inhibited at a molar ratio of 1:1 and chymotrypsin and plasmin to lesser extent

Also inhibits other serine proteases. Has an activity greater than 7,000 BAEE units per mg. A 0.5mg/mL Soybean Trypsin Inhibitor stock solution should be made by dissolving nonsterile powder in 0.01M Borate Buffer or Dulbecco's Phosphate Buffered Saline (DPBS), without calcium and magnesium. Filter sterilize the stock solution and store for up to a week at 2 - 8°C.

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: -5 to -30°C
Shipping conditions: Frozen
Cell Type Mammalian Cells
Form Frozen, powder
Product Type Trypsin inhibitor
Sterility Non-sterile
Without Additives No Calcium, No Magnesium
Quantity 1 g
Shipping Condition Dry Ice
Is Soybean Trypsin Inhibitor a Kunitz-type inhibitor or Bowman-Birk inhibitor?

This product is derived from a Kunitz purification, but in powder form

What reagents do you offer for cell dissociation, and what are the main differences between them?

Please use this selection chart that compares our cell dissociation reagents (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/reagents/trypsin.html).

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.