Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Newborn Calf Serum, New Zealand origin
GREENER_CHOICE

Catalog No. 16010142
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-010-142 1 L
16-010-159 500 mL
16-010-167 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 16-010-142 Supplier Gibco™ Supplier No. 16010142
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Gibco sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

  • Sera Category: Secure
  • Origin: New Zealand
  • Collected from calves 20 days old or less.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Species Cattle/Bovine
Age Newborn (20 days or less)
Country of Origin New Zealand
Form Liquid
Format Bottle
Product Type Newborn Calf Bovine Serum
Quantity 1 L
Serum Treatment Standard (Sterile-filtered)
Sterility Sterile
Shipping Condition Frozen
Show More Show Less
Will improper storage of my tissue culture reagents affect the growth rate of my culture?

Yes. Store animal sera at -5 to -20 degrees C. Store media at 2 to 8 degrees C; use within recommended shelf life period. Store complete media (supplemented) at 2 to 8 degrees C, and for complete medium the recommended shelf life is 2 to 4 weeks. Additionally, minimize exposure of sera and media to light.

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.