Learn More
New England Biolabs, Inc. Gibson Assembly Master Mix – 50 reactions
Supplier: New England Biolabs, Inc. E2611L
Gibson Assembly allows for successful assembly of multiple DNA fragments, regardless of fragment length or end compatibility. Gibson Assembly efficiently joins multiple overlapping DNA fragments in a single-tube isothermal reaction. The Gibson Assembly Master Mix includes three different enzymatic activities that perform in a single buffer: The exonuclease creates single-stranded 3 overhangs that facilitate the annealing of fragments that share complementarity at one end (overlap region).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.