Learn More
New England Biolabs, Inc. Gibson Assembly® Master Mix – 10 reactions
Supplier: New England Biolabs, Inc. E2611S
Gibson Assembly allows for successful assembly of multiple DNA fragments, regardless of fragment length or end compatibility. Gibson Assembly efficiently joins multiple overlapping DNA fragments in a single-tube isothermal reaction. The Gibson Assembly Master Mix includes three different enzymatic activities that perform in a single buffer: The exonuclease creates single-stranded 3 overhangs that facilitate the annealing of fragments that share complementarity at one end (overlap region).
- Assembly and transformation in just under two hours
- Flexible sequence design (scar-less cloning)
- No PCR clean-up step required
- High transformation efficiencies for inserts up to 20 kb
- Easily adapted for multiple DNA manipulations, including site-directed mutagenesis