Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibson Bioscience QC Controls: VRE Swab

Catalog No. p-4882562
Change view
Click to view available options
Species:
Enterococcus faecalis
Enterococcus faecium
Format:
Negative Control
Positive Control
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species Format
21-200-508 Enterococcus faecium Positive Control
21-200-509 Enterococcus faecalis Negative Control
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-200-508 Supplier Gibson Bioscience Supplier No. CE VRE01LEX

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.