Learn More
Gibson Bioscience Reagents: KOH 10%
Specifications
Specifications
| Kit Size | 50mL |
| Product Type | KOH |
| Formulation | Potassium hydroxide 100g, deionized water 1L |
| For Use With (Application) | Demonstration of yeast and fungal elements in wet clinical preparations |
| Storage Requirements | 2° to 25°C |
| Volume (Metric) | 50mL |
| Test Time | 10 min. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.