Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibson Bioscience India Ink Control Kit

Catalog No. 21200402
Change view
Click to view available options
Product Type:
India Ink Negative Control Kit
India Ink Positive Control Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
21-200-402 India Ink Positive Control Kit
21-200-403 India Ink Negative Control Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-200-402 Supplier Gibson Bioscience Supplier No. 100111LEX
Add to Cart
Add to Cart

Requires

The usual microbiological equipment such as slides, coverslips, microscope, biohazard bags, etc. are needed for procedures involving the use of this product.

This kit is utilized as a quality control procedure for India Ink wet mounts

India Ink wet mounts are used for the visualization of encapsulated microorganisms, especially Cryptococcus neoformans.

  • India Ink is added to each positive and negative suspension on a glass slide and observed under a microscope.
  • The positive control provides an encapsulated suspension of Cryptococcus neoformans to be utilized when performing India Ink procedures
  • The negative control provides a non-encapsulated suspension of Candida sp. to be utilized when performing India Ink
EXPECTED RESULTS
  • Positive: The polysaccharide capsules will exclude the particles of ink, and the capsules will appear as clear halos around the organisms on semiopaque backgrounds
  • Negative: No capsules present, no clear halos observed

Specifications

Quantity 20 Tests
Format Dropper
For Use With (Application) Visualization of encapsulated microorganisms
Product Type India Ink Positive Control Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.