Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GIDRP88 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987229
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7229 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7229 Supplier Bioss Supplier No. BS6384R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

R3HCC1L gene ontology (GO) annotation includes protein binding and exon-exon junction complex.

Specifications

Antigen GIDRP88
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene R3HCC1L
Gene Accession No. Q7Z5L2
Gene Alias 1700036B12Rik; AI450607; C10orf28; Coiled-coil domain-containing protein R3HCC1L; D19Ertd386e; GIDRP86; Gidrp88; growth inhibition and differentiation related protein 86; Growth inhibition and differentiation-related protein 88; Growth inhibition and differentiation-related protein 88 homolog; PSORT; Putative mitochondrial space protein 32.1; R3H and coiled-coil domain-containing protein 1-like; R3H domain and coiled-coil containing 1 like; R3H domain and coiled-coil containing 1-like; R3hcc1l; RGD1307934
Gene Symbols R3HCC1L
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GIDRP88.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27291
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.