Learn More
StatLab GIEMSA/MAY-GRUNWALD BONE MARRO
SDP
Supplier:  StatLab KTGMGPT
Encompass_Preferred
This kit stains bone marrow, core sections, mast cells, and Helicobacter pylori in 25 minutes. Stain kit includes Giemsa Stain Stock Pint, May-Grunwald Stain Stock Pint, and 6.8pH Phosphate Buffer Pint.
Catalog No. NC9612699
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.