Learn More
Giemsa's azur-eosin-methylene blue solution, For Microscopy, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Chemical Name or Material | Giemsa's azur-eosin-methylene blue solution |
| UN Number | UN 1230 |
| Odor | Smells like methanol |
| Packaging | Plastic bottle |
| Color | Blue |
| Quantity | 2.5 L |
| Physical Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.