Learn More
Medchemexpress LLC HY-N2523 5mg Medchemexpress, Gigantol CAS:67884-30-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN25235MG
Encompass
Medchemexpress, HY-N2523 5mg Gigantol CAS:67884-30-4 Gigantol is a bibenzyl compound derived from several medicinal orchids. Giganto shows promising therapeutic potential against cancer cells. Gigantol is a novel inhibitor of the Wnt/β-catenin pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3930