Learn More
Sensidyne™ Gilian™ Gilibrator-2™ Spare Parts and Accessories
SDP
Catalog No. 19819036 Shop All Sensidyne Inc. Products
Encompass
Change view
Click to view available options
Product Type:
Standard Flow Sensor and Cell Assembly
Catalog No. Product Type
19-819-036 Standard Flow Sensor and Cell Assembly
1 options
Catalog No. 19-819-036 Supplier Sensidyne™ Supplier No. 8002661
Specifications
Product Type Standard Flow Sensor and Cell Assembly
For Use With (Equipment) Gilibrator-2
Airflow Rate 20 cc/min. to 6 L/min.
Includes Calibration Certificate
Certifications/Compliance NIOSH, NIST