Learn More
StatLab Gill’s Hematoxylin I, 1 Liter
SDP
Supplier:  StatLab HXGHE1LT
Encompass_Preferred
Gill's I is the lightest of three Gill’s Hematoxylin formulations and is best suited for staining in cytology preparations.
Catalog No. NC9901347
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.