Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Gilteritinib
GREENER_CHOICESDP

Catalog No. 789150
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
78-915-0 50 mg
78-911-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 78-915-0 Supplier Tocris Bioscience™ Supplier No. 7891/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent FLT3 inhibitor; also potently inhibits AXL

Gilteritinib is a potent Fms-like tyrosine kinase 3 (FLT3) and AXL inhibitor (IC50 = 0.29 nM and 0.73 nM, for FLT3 and AXL, respectively). In vivo, Gilteritinib reduces tumor growth and improves survival in xenograft and intra-bone marrow transplantation models of FLT3 mutated acute myeloid leukemia. Gilteritinib is orally bioavailable.

Specifications

Chemical Name or Material 6-Ethyl-3-[[3-methoxy-4-[4-(4-methyl-1-piperazinyl)-1-piperidinyl]phenyl]amino]-5-[(tetrahydro-2H-pyran-4-yl)amino]-2-pyrazinecarboxamide
CAS 1254053-43-4
Quantity 50 mg
Target Flt-3/Flk-2/CD135
Molecular Formula C29H44N8O3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.