Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GIMAP2 Monoclonal Antibody (1E10)
GREENER_CHOICE

Catalog No. PIMA521620
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521620 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521620 Supplier Invitrogen™ Supplier No. MA521620
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MDQNEHSHWG PHAKGQCASR SELRIILVGK TGTGKSAAGN SILRKQAFES KLGSQTLTKT CSKSQGSWGN REIVIIDTPD MFSWKDHCEA LYKEVQRCYL.

This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. In humans, the IAN subfamily genes are located in a cluster at 7q36.1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIMAP2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1E10
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene GIMAP2
Gene Accession No. Q9UG22
Gene Alias D15S9; GIMAP2; GTPase IMAP family member 2; GTPase, IMAP family member 2; HIMAP2; IAN12; IMAP2; immune-associated nucleotide-binding protein 12; immunity associated protein 2; immunity-associated protein 2; RNF63; ZNF127
Gene Symbols GIMAP2
Host Species Mouse
Immunogen GIMAP2 (NP_056475.1, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26157
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.