Learn More
TARGETMOL CHEMICALS INC GINKGETIN 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T4S21265MG
Encompass_Preferred
Also available in 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Ginkgetin has anti-influenza virus and anti-fungal activities. 2. Ginkgetin has anti-inflammatory activity can down-regulates COX-2 induction in vivo against skin inflammatory responses. 3. Ginkgetin is a good STAT3 inhibitor and may be a useful lead molecule for development of a therapeutic STAT3 inhibitor. 4. Ginkgetin induces apoptosis in PC-3 cells via activation of caspase 3 and inhibition of survival genes as a potent chemotherapeutic agent for prostate cancer treatment. 5. Ginkgetin has neuroprotective activity against neurological injury induced by MPTP occurs via regulating iron homeostasis it may provide neuroprotective therapy for Parkinson's disease and iron metabolism disorder related diseases. purity: 99%
Catalog No. 50-257-6715
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.