Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ginkgolide B from Ginkgo biloba leaves, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013219 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3219 10 mg
11-101-3220 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-3219 Supplier MilliporeSigma Supelco Supplier No. G691010MG
Add to Cart
Add to Cart

Ginkgolide B from Ginkgo biloba leaves is a terpene lactone and one of the main molecular form of ginkgolides, present in Ginkgo biloba leaves. It is widely used to treat peripheral and cerebral circulation disorders.

Food and Beverages
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD05662347
Synonym BN-52021

Specifications

CAS 15291-77-7
Packaging Glass Insert
Linear Formula C20H24O10
Quantity 10 mg
Formula Weight 424.40
Percent Purity ≥90% (HPLC)
Physical Form Neat
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.