Learn More
Medchemexpress LLC Ginkgolide J | 107438-79-9 | MFCD03093737 | 100.0% | 424.40 g·mol⁻¹ | C20H24O10 | 10 MG
Supplier: Medchemexpress LLC HYN078610MG
Ginkgolide J is a naturally occurring diterpenoid from Ginkgo biloba used in research for its reported neuroprotective and anti-apoptotic activity (IC50 12-54 μM). It is supplied as a high-purity powder intended for laboratory use, with solubility information and handling documentation provided for in vitro and in vivo formulation.
- Natural diterpenoid with neuroprotective activity.
- Reported IC50 range of 12-54 μM in published assays.
- High purity suitable for research applications.
- Soluble in DMSO; in vivo formulation protocols available.
- Provided in mg-scale packs for small-scale experiments.
- Documentation includes data sheet, COA, and SDS for handling and quality confirmation.