Learn More
Cayman Chemical PrednIcarbate 10mg
SDP
Supplier:  Cayman Chemical 2366810 MG
Encompass_Preferred
An ionophore antibiotic used in veterinary practice as a coccidiostat for gastrointestinal parasites inhibits NF-KB signaling via inhibition of IKBa phosphorylation (IC50 3 2 UM) and stimulates TRAIL-mediated apoptosis in glioma cells via ER stress
Catalog No. 50-299-4107
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.