Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ginsenoside Rg1, Thermo Scientific™

Catalog No. AC469151000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469151000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC469151000 Supplier Thermo Scientific Chemicals Supplier No. 469151000
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 22427-39-0
Molecular Formula C42 H72 O14
Molecular Weight (g/mol) 801.02
MDL Number MFCD00210293
InChI Key YURJSTAIMNSZAE-HHNZYBFYSA-N
IUPAC Name (2S,3R,4S,5S,6R)-2-{[(2S)-2-[(1S,3aR,3bR,5S,5aR,7S,9aR,9bR,11R,11aR)-7,11-dihydroxy-3a,3b,6,6,9a-pentamethyl-5-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]-6-methylhept-5-en-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CC(C)=CCC[C@](C)(O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@H]1CC[C@]2(C)[C@@H]1[C@H](O)C[C@@H]1[C@@]3(C)CC[C@H](O)C(C)(C)[C@@H]3[C@H](C[C@@]21C)O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

Specifications

Quantity 100 mg
Melting Point 188°C to 191°C
Formula Weight 801.02
Physical Form Crystalline Powder or Crystals
Color Colorless or White
Purity ≥90%
Chemical Name or Material Ginsenoside Rg1
Hazard Category Exclamation mark
EINECSNumber 244-989-9
Recommended Storage Normal conditions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.