Learn More
Invitrogen™ GIPC1 Polyclonal Antibody

Description
Immunogen sequence: MPLGLGRRKK APPLVENEEA EPGRG Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 92%.
Specifications
Specifications
| Antigen | GIPC1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.10 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | GIPC1 |
| Gene Accession No. | O14908 |
| Gene Alias | C19orf3; GAIP C-terminus-interacting protein; Gipc; GIPC PDZ domain containing family member 1; GIPC PDZ domain containing family, member 1; Gipc1; GLUT1 C-terminal binding protein; GLUT1 C-terminal-binding protein; GLUT1CBP; Glut1CIP; Hs.6454; IGF-1 receptor interacting protein 1; IIP-1; neurophilin1-IP; NIP; PDZ domain-containing protein GIPC1; regulator of G-protein signaling 19 interacting protein 1; regulator of G-protein signalling 19 interacting protein 1; Rgs19; RGS19-interacting protein 1; RGS19IP1; RGS-GAIP interacting protein C-terminus; RGS-GAIP interacting protein GIPC; RGS-GAIP interacting protein, C-terminus; RGS-GAIP-interacting protein; semaF cytoplasmic domain associated protein; semaF cytoplasmic domain-associated protein 1; SEMCAP; Semcap1; SEMCAP-1; SYNECTIIN; SYNECTIN; Tax interacting protein, clone 2; tax interaction protein 2; TaxIP2; TIP-2 |
| Gene Symbols | GIPC1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.