Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GeneBLAzer™ GeneBLAzer™ GIPR-CRE-bla HEK 293T Cells

Catalog No. K1864
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1864 1 vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1864 Supplier GeneBLAzer™ Supplier No. K1864
Add to Cart
Add to Cart
Request a Quote

GeneBLAzer™ GIPR-CRE-bla HEK 293T Cells contain the human Gastric Inhibitory Peptide Receptor (GIPR).

  • Accession Number: NM_000164.2
  • Population Status: Single Cell Clone Origin
  • GeneBLAzer™ GIPR-CRE-bla HEK 293T cells contain the human Gastric Inhibitory Peptide Receptor (GIPR), stably integrated into the CellSensor™ CRE-bla HEK 293T cell line. CellSensor™ CRE-bla HEK 293T cells (Cat. no. K1535)) contain a beta-lactamase reporter gene under control of the CRE Response Element. The GeneBLAzer™ GIPR-CRE-bla HEK 293T cells are functionally validated for Z’-factor and EC50 concentrations of Gastric Inhibitory Peptide (GIP). In addition, GeneBLAzer™ GIPR-CRE-bla HEK 293T cells have been tested for assay performance under variable conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_000164.2
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage GeneBLAzer™ GIPR-CRE-bla HEK 293T cells are shipped on dry ice, 8 x 10^6 cells⁄vial. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Detection Method Fluorescence
For Use With (Application) FRET
Human Protein Class G-Protein Coupled Receptor (GPCR)
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 1 vial
Readout End Point
Target Entry GIPR
Type Cell Line
Cell Line 293T (HEK)
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.