Learn More
Description
Specifications
Specifications
| Antigen | GITR Ligand |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MIH44 |
| Concentration | 0.2 mg/mL |
| Conjugate | RY586 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | Tnfsf18; Gitrl Glucocorticoid-induced-tumor necrosis factor receptor ligand |
| Host Species | Rat |
| Immunogen | Mouse GITRL Transfected Cell Line |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.