Learn More
Invitrogen™ GKN1 Polyclonal Antibody

Description
The amino acid sequence used as immunogen is 100% homologous in chimpanzee and dog, 94% homologous in mouse and cow, 88% homologous in rat and 77% homologous in chicken.
Specifications
Specifications
| Antigen | GKN1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | GKN1 |
| Gene Accession No. | Q8HYA9, Q9NS71 |
| Gene Alias | 18 kDa antrum mucosa protein; 2200002K21Rik; Amp18; AMP-18; Amp18; gastrokine; BRICD1; BRICHOS domain containing 1; Ca11; FOV; foveolin; gastrokine 1; gastrokine-1; Gkn1; Protein CA11; Protein CA11 homolog; UNQ489/PRO1005 |
| Gene Symbols | GKN1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.