Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transforming Technologies GL6500N - ESD Level 6 Cut Resistant Gloves - Nitrile Palm Coated
SDP

Catalog No. 17226985
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-226-985 X-Small
17-226-986 Small
17-226-988 Medium
17-226-989 Large
17-226-990 X-Large
17-226-991 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-226-985 Supplier Transforming Technologies Supplier No. GL6501N
Add to Cart
Add to Cart

The StaticCare ESD Level 6 Cut Resistant Gloves are ANSI Cut Level A6 Certified.

  • Sold as 12 pair per bag.
  • Applications:
  • Automotive industry
  • Glass industry
  • Paper industry
  • Construction
  • Electronics Industry

Specifications

Coated Area Palm
Coating Material Nitrile
Color Gray, Black
Hand Ambidextrous
Material Composite Yarn
Product Type Cut-resistant Gloves
Size X-Small
Gender Unisex
Glove Cuff Style Knitwrist
Packaging Type Box
Quantity 12 pack
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.