Learn More
Glas Col LLC Heating Mantle
SDP
Supplier:  Glas Col LLC 100A MO1083
Encompass_Preferred
Fabric heating top for 1L flask. 115V heat to 400 degrees C internal operating temperature. This mantle helps prevent vapor condensation in the upper half of a flask during distillation of high-boiling materials. The tops are designed for use with Series M, TM, or STM mantles. They protect the flask, the solutions, and the laboratory from drafts, which would disturb the reflux ration during fractionation, from flying glass of flask implosions in high vacuum distillations, from falling objects; and, from thermal strain between top and lower half of flask during high-temperature operations. With a glass-fabric construction and laced-side opening, the elliptical hole accomodates 1-, 2-, and 3-neck flasks.All 115 volt units are CSA certified.
Catalog No. 50-161-6351
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.