Learn More
Eisco™ Glass Adapter Receivers
SCI_ED
Catalog No. S88398 Shop All Eisco Products
Change view
Click to view available options
Standard Taper Joint No.:
19/26
Catalog No. Standard Taper Joint No.
S88398 19/26
1 options
Catalog No. S88398 Supplier Eisco™ Supplier No. CH01018B
Add to Cart
Add to Cart
Eisco™ Glass Adapter Receivers with screw threads to provide additional safety in lab.
  • With screw threads
  • Made from high quality Borosilicate 3.3 Glass
  • Screw threads provide additional lab safety
  • Screw thread joints allow flasks to connect firmly with other glassware
  • Autoclavable
Specifications
For Use With (Application) Distillation set ups
Height (Metric) 9 cm
Length (Metric) 3 cm
Material Borosilicate Glass (3.3)
Product Type Receiver
Quantity 1
Standard Taper Joint No. 19/26
Width (Metric) 3 cm