Learn More
BUCHI Glass Caps
SDP
Catalog No. 05405650 Shop All BUCHI Products
Change view
Click to view available options
For Use With (Equipment):
Digestion Units
Catalog No. For Use With (Equipment)
05-405-650 Digestion Units
1 options
Catalog No. 05-405-650 Supplier BUCHI Supplier No. 040049
Add to Cart
Add to Cart
For occupying empty sample positions, to close the suction circuit.
  • For use on all BUCHI Digestion Units
Specifications
Product Type Cap
For Use With (Application) Sealing the Suction Circuit
For Use With (Equipment) Digestion Units
Material Glass