Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Glass Crow Receivers
SCI_ED

Catalog No. S88808 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S88808 50 mL
S88807 25 mL
S88809 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S88808 Supplier Eisco™ Supplier No. CH0351B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High quality, Class B, borosilicate 3.3 glass crow receivers. Perfect for distillates in oil and tar tests.

  • Crow receivers made from high quality borosilicate 3.3 glass
  • Class B, graduated
  • Perfect for distillates in oil and tar tests
  • Warranty and Services

    1 year limited manufacturer warranty

    Specifications

    Material Borosilicate Glass (3.3)
    Capacity (Metric) 50 mL
    Graduations 0.5 mL
    Capacity (English) 1.7 oz.
    Class Class A
    Tolerance Strain Point -515°C, Annealing Point 565°C, Softening Point 820°C
    Product Type Graduated Cylinder
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.