Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Glass Equilateral Prism
SCI_ED

Catalog No. S27715 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S27715 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27715 Supplier Eisco™ Supplier No. PH0556C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glass Equilateral Prism is ideal for physical science demonstration and experiments

  • Non optically worked glass
  • Slightly beveled edges
  • Side Length: 50 mm
  • Angles: 60°

Specifications

Product Type Prism, Equilateral
Description 60° Angles
Material Glass
For Use With (Application) Physical Science Demonstrations and Experiments
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.