Learn More
Eisco™ Glass Equilibrium Tube Set
SCI_ED
Catalog No. S27648 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
S27648 1 Ea.
1 options
Catalog No. S27648 Supplier Eisco™ Supplier No. PH0201A
Add to Cart
Add to Cart
This glass equilibrium tube set includes 4 borosilicate glass tubes of different shapes to illustrate that liquids will maintain the same level, regardless of shape
  • Includes 4 borosilicate glass tubes of different shapes
  • Mounted on a plastic base
  • 6.25 in. tall
Specifications
Product Type Equilibrium Tube Set
Material Borosilicate Glass
For Use With (Application) Equilibrium Demonstrations
Includes 4 Tubes, Plastic Mounting Base
Quantity 1 Ea.
Height (English) 6.25 in.